4-Chloro-2-propoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

273180-1G 273180-500MG 273180-5G 273180-25G
77467-924EA 260.59 USD
77467-924 77480-620 77490-968 77501-022
4-Chloro-2-propoxybenzaldehyde ≥95%
4-Chloro-2-propoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34880173
CAS Number: 1595953-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR