1-[1,1'-Biphenyl]-3-yl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

271923-1G 271923-500MG 271923-5G
77467-920EA 471.74 USD
77467-920 77480-616 77490-962
1-[1,1'-Biphenyl]-3-yl-1-propanone ≥95%
1-[1,1'-Biphenyl]-3-yl-1-propanone

Precaución: For research use only

Formula: C₁₅H₁₄O
MW: 210.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09042444
CAS Number: 107075-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR