Benzo[b]thiophene-2-carboxylic acid, 3-methyl-, ethyl ester ≥95%

Supplier: AOBChem USA

Warning

271483-1G 271483-500MG 271483-5G 271483-25G
77467-916EA 347.11 USD
77467-916 77480-612 77490-956 77501-012
Benzo[b]thiophene-2-carboxylic acid, 3-methyl-, ethyl ester ≥95%
Benzo[b]thiophene-2-carboxylic acid, 3-methyl-, ethyl ester

Precaución: For research use only

Formula: C₁₂H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD06204391
CAS Number: 31310-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR