1-(2-Dibenzofuranyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

270539-1G 270539-500MG 270539-5G 270539-25G
77467-908EA 415.09 USD
77467-908 77480-604 77490-946 77501-002
1-(2-Dibenzofuranyl)-2,2-difluoroethanone ≥95%
1-(2-Dibenzofuranyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₄H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34879061
CAS Number: 1979947-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR