(3-Bromobenzo[b]thien-2-yl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

270505-1G 270505-500MG 270505-5G 270505-25G
77467-904EA 415.09 USD
77467-904 77480-600 77490-942 77500-998
(3-Bromobenzo[b]thien-2-yl)phenylmethanone ≥95%
(3-Bromobenzo[b]thien-2-yl)phenylmethanone

Precaución: For research use only

Formula: C₁₅H₉BrOS
MW: 317.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34879050
CAS Number: 99661-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR