2,2,2-Trifluoro-1-(5-methoxy-2-thienyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

270485-1G 270485-500MG 270485-5G
77467-900EA 546.93 USD
77467-900 77480-596 77490-938
2,2,2-Trifluoro-1-(5-methoxy-2-thienyl)ethanone ≥95%
2,2,2-Trifluoro-1-(5-methoxy-2-thienyl)ethanone

Precaución: For research use only

Formula: C₇H₅F₃O₂S
MW: 210.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30471181
CAS Number: 24124-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR