4-Ethyl-2-(ethylthio)-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

270194-1G 270194-500MG 270194-5G 270194-25G
77467-896EA 415.09 USD
77467-896 77480-592 77490-934 77500-990
4-Ethyl-2-(ethylthio)-1-methoxybenzene ≥95%
4-Ethyl-2-(ethylthio)-1-methoxybenzene

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22484332
CAS Number: 1824367-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR