3-Bromo-2,6-difluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

269983-1G 269983-500MG 269983-5G 269983-25G
77467-892EA 415.09 USD
77467-892 77480-588 77490-930 77500-986
3-Bromo-2,6-difluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥95%
3-Bromo-2,6-difluoro-α-methyl-α-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34878856
CAS Number: 1700379-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR