2-Fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

269947-1G 269947-500MG 269947-5G 269947-25G
77467-888EA 415.09 USD
77467-888 77480-584 77490-926 77500-982
2-Fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥95%
2-Fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068027
CAS Number: 1158038-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR