5-Bromo-2-fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

269896-1G 269896-500MG 269896-5G 269896-25G
77467-884EA 255.44 USD
77467-884 77480-580 77490-922 77500-978
5-Bromo-2-fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥97%
5-Bromo-2-fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34878812
CAS Number: 1598357-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR