Ethyl 4-(4-chlorophenyl)-4-hydroxy-1-piperidinecarboxylate ≥95%
Supplier: AOBChem USA
|
Warning
|
269487-1G
269487-500MG
269487-5G
269487-25G
77467-880EA
415.09
USD
77467-880
77480-576
77490-918
77500-976
Ethyl 4-(4-chlorophenyl)-4-hydroxy-1-piperidinecarboxylate ≥95%
Ethyl 4-(4-chlorophenyl)-4-hydroxy-1-piperidinecarboxylate
Precaución: For research use only
Formula:
C₁₄H₁₈ClNO₃ MW: 283.75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34878655 CAS Number: 23482-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...