Cyclopropyl(2-fluoro-5-(trifluoromethyl)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

269400-1G 269400-500MG 269400-5G
77467-876EA 646.84 USD
77467-876 77480-574 77490-914
Cyclopropyl(2-fluoro-5-(trifluoromethyl)phenyl)methanol ≥95%
Cyclopropyl(2-fluoro-5-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₀F₄O
Storage Temperature: Ambient
MDL Number: MFCD23823071
CAS Number: 1516844-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR