3-Cyclohexen-1-ol, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1-benzoate ≥97%

Supplier: AOBChem USA

Warning

26921-1G 26921-500MG
77467-872EA 1447.15 USD
77467-872 77480-570
3-Cyclohexen-1-ol, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1-benzoate ≥97%
3-Cyclohexen-1-ol, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1-benzoate

Precaución: For research use only

Formula: C₁₉H₂₅BO₄
MW: 328.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756688
CAS Number: 2121513-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR