1-Fluoro-4-[(4-nitrophenyl)sulfanyl]benzene ≥96%

Supplier: AOBChem USA

Warning

Synonyms: (4-Fluorophenyl)(4-nitrophenyl)sulphane, 4-[(4-Fluorophenyl)thio]nitrobenzene

269113-1G 269113-500MG 269113-5G 269113-25G
77467-868EA 107.12 USD
77467-868 77480-566 77490-908 77500-968
1-Fluoro-4-[(4-nitrophenyl)sulfanyl]benzene ≥96%
1-Fluoro-4-[(4-nitrophenyl)sulfanyl]benzene

Precaución: For research use only

Formula: C₁₂H₈FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD04117815
CAS Number: 2438-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR