3-(2-Fluorophenyl)tetrahydro-3-furanol ≥97%

Supplier: AOBChem USA

Warning

268489-1G 268489-500MG 268489-5G 268489-25G
77467-852EA 255.44 USD
77467-852 77480-552 77490-892 77500-954
3-(2-Fluorophenyl)tetrahydro-3-furanol ≥97%
3-(2-Fluorophenyl)tetrahydro-3-furanol

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18332736
CAS Number: 1339589-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR