Tetrahydro-3-phenyl-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268470-1G 268470-500MG 268470-5G
77467-848EA 447.02 USD
77467-848 77480-548 77490-888
Tetrahydro-3-phenyl-3-furanol ≥95%
Tetrahydro-3-phenyl-3-furanol

Precaución: For research use only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18332781
CAS Number: 102654-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR