Tetrahydro-3-[4-(2-methylpropyl)phenyl]-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268393-1G 268393-500MG 268393-5G 268393-25G
77467-844EA 415.09 USD
77467-844 77480-544 77490-884 77500-948
Tetrahydro-3-[4-(2-methylpropyl)phenyl]-3-furanol ≥95%
Tetrahydro-3-[4-(2-methylpropyl)phenyl]-3-furanol

Precaución: For research use only

Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31694030
CAS Number: 2707492-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR