α-Butyl-3-fluoro-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

268159-1G 268159-500MG 268159-5G 268159-25G
77467-840EA 260.59 USD
77467-840 77480-540 77490-880 77500-944
α-Butyl-3-fluoro-α-methylbenzenemethanol ≥95%
α-Butyl-3-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
Storage Temperature: Ambient
MDL Number: MFCD21128215
CAS Number: 1494629-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR