3-Bromo-α-butyl-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

268086-1G 268086-500MG 268086-5G 268086-25G
77467-836EA 297.67 USD
77467-836 77480-536 77490-876 77500-940
3-Bromo-α-butyl-α-methylbenzenemethanol ≥97%
3-Bromo-α-butyl-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21140617
CAS Number: 1149357-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR