N-(2-Furanylmethyl)benzamide ≥95%

Supplier: AOBChem USA

Warning

267563-1G 267563-500MG 267563-5G 267563-25G
77467-832EA 305.91 USD
77467-832 77480-532 77490-872 77500-936
N-(2-Furanylmethyl)benzamide ≥95%
N-(2-Furanylmethyl)benzamide

Precaución: For research use only

Formula: C₁₂H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00453779
CAS Number: 3952-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR