4-Bromo-3-cyclohexen-1-ol ≥97%

Supplier: AOBChem USA

Warning

26751-1G 26751-500MG 26751-5G 26751-25G
77467-828EA 415.09 USD
77467-828 77480-528 77490-868 77500-934
4-Bromo-3-cyclohexen-1-ol ≥97%
4-Bromo-3-cyclohexen-1-ol

Precaución: For research use only

Formula: C₆H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD29949914
CAS Number: 1450812-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR