1-(5-Bromo-2-fluorophenyl)-1,2-ethanediol ≥97%

Supplier: AOBChem USA

Warning

267140-1G 267140-500MG 267140-5G
77467-824EA 810.61 USD
77467-824 77480-524 77490-864
1-(5-Bromo-2-fluorophenyl)-1,2-ethanediol ≥97%
1-(5-Bromo-2-fluorophenyl)-1,2-ethanediol

Precaución: For research use only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
CAS Number: 1555837-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR