1-(2,4-Dichlorophenyl)-1,2-ethanediol ≥95%

Supplier: AOBChem USA

Warning

266976-1G 266976-500MG 266976-5G
77467-820EA 457.32 USD
77467-820 77480-520 77490-860
1-(2,4-Dichlorophenyl)-1,2-ethanediol ≥95%
1-(2,4-Dichlorophenyl)-1,2-ethanediol

Precaución: For research use only

Formula: C₈H₈Cl₂O₂
Storage Temperature: Ambient
CAS Number: 14866-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR