1-(2,5-Dichlorophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

266869-1G 266869-500MG 266869-5G 266869-25G
77467-816EA 415.09 USD
77467-816 77480-516 77490-856 77500-928
1-(2,5-Dichlorophenyl)-2-methylpropan-1-ol ≥97%
1-(2,5-Dichlorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂Cl₂O
MW: 219.1076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787835
CAS Number: 1247161-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR