2-(2-Fluoro-5-(trifluoromethyl)phenyl)tetrahydro-2H-pyran-2-ol ≥95%

Supplier: AOBChem USA

Warning

266847-1G 266847-500MG 266847-5G 266847-25G
77467-812EA 415.09 USD
77467-812 77480-512 77490-852 77500-924
2-(2-Fluoro-5-(trifluoromethyl)phenyl)tetrahydro-2H-pyran-2-ol ≥95%
2-(2-Fluoro-5-(trifluoromethyl)phenyl)tetrahydro-2H-pyran-2-ol

Precaución: For research use only

Formula: C₁₂H₁₂F₄O₂
MW: 264.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34877796
CAS Number: 2969473-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR