1-[3,5-Dimethyl-4-(1-methylethoxy)phenyl]ethanone ≥97%

Supplier: AOBChem USA

Warning

266486-1G 266486-500MG 266486-5G
77467-804EA 572.68 USD
77467-804 77480-504 77490-844
1-[3,5-Dimethyl-4-(1-methylethoxy)phenyl]ethanone ≥97%
1-[3,5-Dimethyl-4-(1-methylethoxy)phenyl]ethanone

Precaución: For research use only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD13813473
CAS Number: 1119441-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR