9,9'-Spirobi[9H-fluorene]-2-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

266365-1G 266365-500MG 266365-5G 266365-25G
77467-800EA 415.09 USD
77467-800 77480-500 77490-840 77500-912
9,9'-Spirobi[9H-fluorene]-2-carboxaldehyde ≥95%
9,9'-Spirobi[9H-fluorene]-2-carboxaldehyde

Precaución: For research use only

Formula: C₂₆H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD30532219
CAS Number: 124575-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR