4'-Bromo-2,6-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

26587-1G 26587-500MG 26587-5G 26587-25G
77467-796EA 224.54 USD
77467-796 77480-496 77490-836 77500-910
4'-Bromo-2,6-dimethoxy-1,1'-biphenyl ≥97%
4'-Bromo-2,6-dimethoxy-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₄H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD27756669
CAS Number: 168849-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR