1-(4-Bromophenyl)ethane-1,2-diol ≥97%

Supplier: AOBChem USA

Warning

264898-1G 264898-500MG 264898-5G 264898-25G
77467-792EA 162.74 USD
77467-792 77480-492 77490-832 77500-904
1-(4-Bromophenyl)ethane-1,2-diol ≥97%
1-(4-Bromophenyl)ethane-1,2-diol

Precaución: For research use only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743652
CAS Number: 92093-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR