Triethyl(2,4,6-Trifluorophenyl)silane ≥97%

Supplier: AOBChem USA

Warning

264888-1G 264888-500MG 264888-5G 264888-25G
77467-788EA 415.09 USD
77467-788 77480-488 77490-828 77500-902
Triethyl(2,4,6-Trifluorophenyl)silane ≥97%
Triethyl(2,4,6-Trifluorophenyl)silane

Precaución: For research use only

Formula: C₁₂H₁₇F₃Si
MW: 246.3440 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34877177
CAS Number: 2969435-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR