(2,6-Difluoro-3-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

264886-1G 264886-500MG 264886-5G 264886-25G
77467-784EA 415.09 USD
77467-784 77480-484 77490-824 77500-898
(2,6-Difluoro-3-methylphenyl)triethylsilane ≥95%
(2,6-Difluoro-3-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀F₂Si
Storage Temperature: Ambient
MDL Number: MFCD34877175
CAS Number: 2969242-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR