1-(2-Fluoro-5-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264582-1G 264582-500MG 264582-5G 264582-25G
77467-780EA 394.49 USD
77467-780 77480-480 77490-818 77500-892
1-(2-Fluoro-5-methylphenyl)-1-propanone ≥95%
1-(2-Fluoro-5-methylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₁₁FO
MW: 166.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222554
CAS Number: 575-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR