4'-tert-Butylpropiophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-[4-(1,1-Dimethylethyl)phenyl]-1-propanone

264499-1G 264499-5G 264499-25G
77467-772EA 62.83 USD
77467-772 77490-810 77500-886
4'-tert-Butylpropiophenone ≥95%
4'-tert-Butylpropiophenone

Precaución: For research use only

Formula: C₁₃H₁₈O
MW: 190.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100434
CAS Number: 71209-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR