6-Amino-2-methylpyridin-3-ylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26362-1G 26362-500MG 26362-5G 26362-25G
77467-768EA 396.55 USD
77467-768 77480-470 77490-806 77500-882
6-Amino-2-methylpyridin-3-ylboronic acid pinacol ester ≥97%
6-Amino-2-methylpyridin-3-ylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₂H₁₉BN₂O₂
MW: 234.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182009
CAS Number: 1220219-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR