2-[4-(Methylthio)phenyl]pyridine ≥95%

Supplier: AOBChem USA

Warning

263001-1G 263001-500MG 263001-5G 263001-25G
77467-756EA 341.96 USD
77467-756 77480-458 77490-794 77500-870
2-[4-(Methylthio)phenyl]pyridine ≥95%
2-[4-(Methylthio)phenyl]pyridine

Precaución: For research use only

Formula: C₁₂H₁₁NS
MW: 201.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201383
CAS Number: 4373-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR