1-Bromo-3-chloro-5-cyclopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

26287-1G 26287-500MG 26287-5G
77467-752EA 684.95 USD
77467-752 77480-456 77490-790
1-Bromo-3-chloro-5-cyclopropylbenzene ≥95%
1-Bromo-3-chloro-5-cyclopropylbenzene

Precaución: For research use only

Formula: C₉H₈BrCl
Storage Temperature: Ambient
MDL Number: MFCD19439934
CAS Number: 945717-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR