3,5-Dimethyl-1H-pyrazolo[3,4-b]pyridine ≥97%

Supplier: AOBChem USA

Warning

26181-1G 26181-500MG 26181-5G
77467-748EA 772.5 USD
77467-748 77480-452 77490-786
3,5-Dimethyl-1H-pyrazolo[3,4-b]pyridine ≥97%
3,5-Dimethyl-1H-pyrazolo[3,4-b]pyridine

Precaución: For research use only

Formula: C₈H₉N₃
MW: 147.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045261
CAS Number: 1449008-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR