2-Benzyl-4-bromophenol ≥97%

Supplier: AOBChem USA

Warning

26157-1G 26157-500MG 26157-5G 26157-25G
77467-740EA 328.57 USD
77467-740 77480-444 77490-778 77500-858
2-Benzyl-4-bromophenol ≥97%
2-Benzyl-4-bromophenol

Precaución: For research use only

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17677457
CAS Number: 19578-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR