1-(4-Isopropoxyphenyl)propan-1-one ≥96%

Supplier: AOBChem USA

Warning

261392-5G 261392-25G
77490-774EA 255.44 USD
77490-774 77500-854
1-(4-Isopropoxyphenyl)propan-1-one ≥96%
1-(4-Isopropoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD02176649
CAS Number: 35081-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR