1-Bromo-4-(cyclopentyloxy)benzene ≥96%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-(cyclopentyloxy)benzene

260758-5G 260758-25G
77490-750EA 559.29 USD
77490-750 77500-832
1-Bromo-4-(cyclopentyloxy)benzene ≥96%
1-Bromo-4-(cyclopentyloxy)benzene

Precaución: For research use only

Formula: C₁₁H₁₃BrO
MW: 241.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461672
CAS Number: 30752-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR