tert-Butyl N-(3-methylpyridin-4-yl)carbamate ≥97%

Supplier: AOBChem USA

26032-1G 26032-500MG 26032-5G 26032-25G
77467-704EA 245.14 USD
77467-704 77480-412 77490-740 77500-816
tert-Butyl N-(3-methylpyridin-4-yl)carbamate ≥97%
tert-Butyl N-(3-methylpyridin-4-yl)carbamate

Precaución: For research use only

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD11976851
CAS Number: 180253-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR