3-Chloro-α-cyclopropyl-2,6-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

259517-1G 259517-500MG 259517-5G
77467-688EA 522.21 USD
77467-688 77480-396 77490-724
3-Chloro-α-cyclopropyl-2,6-difluorobenzenemethanol ≥95%
3-Chloro-α-cyclopropyl-2,6-difluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₉ClF₂O
MDL Number: MFCD34875835
CAS Number: 2354614-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR