α-Cyclopropyl-2,5-difluorobenzenemethanol ≥97%

Supplier: AOBChem USA

259506-1G 259506-500MG 259506-5G 259506-25G
77467-684EA 226.6 USD
77467-684 77480-392 77490-720 77500-808
α-Cyclopropyl-2,5-difluorobenzenemethanol ≥97%
α-Cyclopropyl-2,5-difluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₀F₂O
MDL Number: MFCD12787112
CAS Number: 1225507-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR