α-Cyclopropyl-2,3-difluoro-4-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

259471-1G 259471-500MG 259471-5G 259471-25G
77467-676EA 340.93 USD
77467-676 77480-384 77490-712 77500-802
α-Cyclopropyl-2,3-difluoro-4-methylbenzenemethanol ≥97%
α-Cyclopropyl-2,3-difluoro-4-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O
MDL Number: MFCD34149019
CAS Number: 1692339-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR