3-Chloro-α-cyclopropyl-2-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

259441-1G 259441-500MG 259441-5G
77467-672EA 497.49 USD
77467-672 77480-380 77490-708
3-Chloro-α-cyclopropyl-2-fluorobenzenemethanol ≥95%
3-Chloro-α-cyclopropyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₀ClFO
MDL Number: MFCD20400974
CAS Number: 1340540-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR