α-Cyclopropyl-4-ethoxy-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

259385-1G 259385-500MG 259385-5G
77467-664EA 437.75 USD
77467-664 77480-372 77490-700
α-Cyclopropyl-4-ethoxy-2-methylbenzenemethanol ≥95%
α-Cyclopropyl-4-ethoxy-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MDL Number: MFCD34875802
CAS Number: 2352680-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR