α-Cyclopropyl-2-ethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

259374-1G 259374-500MG 259374-5G
77467-660EA 447.02 USD
77467-660 77480-368 77490-696
α-Cyclopropyl-2-ethoxybenzenemethanol ≥95%
α-Cyclopropyl-2-ethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MDL Number: MFCD17223686
CAS Number: 1339347-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR