α-Cyclopropyl-3-(1-methylethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

259369-1G 259369-500MG 259369-5G
77467-652EA 477.92 USD
77467-652 77480-360 77490-688
α-Cyclopropyl-3-(1-methylethoxy)benzenemethanol ≥95%
α-Cyclopropyl-3-(1-methylethoxy)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MDL Number: MFCD21328302
CAS Number: 1493199-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR