α-Cyclopropyl-4-(1-methylethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

259333-1G 259333-500MG 259333-5G 259333-25G
77467-644EA 347.11 USD
77467-644 77480-352 77490-680 77500-792
α-Cyclopropyl-4-(1-methylethoxy)benzenemethanol ≥95%
α-Cyclopropyl-4-(1-methylethoxy)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MDL Number: MFCD12787618
CAS Number: 1225504-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR