α-Cyclopropyl-3-fluorobenzenemethanol ≥96%

Supplier: AOBChem USA

259304-1G 259304-500MG 259304-5G 259304-25G
77467-632EA 226.6 USD
77467-632 77480-340 77490-668 77500-786
α-Cyclopropyl-3-fluorobenzenemethanol ≥96%
α-Cyclopropyl-3-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₁FO
MDL Number: MFCD12153720
CAS Number: 844470-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR